EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
Closing the ring to bring up the light: synthesis of a hexacyclic acridinium cyanine dye.
PMID 22907584
Abstract
The synthesis of a geometrically constrained and near-planar hexacyclic acridinium cyanine dye 9 is reported. When compared to its unlocked and non-fluorescent monomethine cyanine dye analogue 3, this photostable dye emits in the green area of the spectrum with a remarkable quantum yield close to unity in organic solvents and above 0.5 in water. A detailed steady-state and time-resolved spectroscopic study revealed that dye 9 forms emissive aggregates in water, which are responsible for a red-shifted and broadened emission band and longer emission lifetime, τ≈33 compared to 6.5-7.0 ns for the monomeric dye. Dye 9 also binds strongly to DNA (both duplex and quadruplex) in its monomeric form and is very efficiently taken up by cells, in which it accumulates primarily into the nucleus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
166510
1,1′-Diethyl-2,2′-carbocyanine iodide, 97%
C25H25IN2
201715
Pinacyanol chloride
C25H25ClN2