EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A diversity-oriented approach to spiroindolines: post-Ugi gold-catalyzed diastereoselective domino cyclization.
PMID 22907654
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I5605
Indoline, ReagentPlus®, 99%
C8H9N