EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
A phenylboronate-functionalized polyion complex micelle for ATP-triggered release of siRNA.
PMID 22907668
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
78181
Phenylboronic acid, purum, ≥97.0% (HPLC)
C6H7BO2
P20009
Phenylboronic acid, 95%
C6H7BO2