EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
A carbazole-containing porphyrinoid: synthesis and oxidation to the porphyrin-state.
PMID 22908132
Abstract
Carbazole-containing porphyrinoid was synthesised for the first time via Suzuki-Miyaura cross-coupling reaction. Oxidation with MnO(2) yielded its porphyrin-state featuring macrocyclic aromaticity in exchange for the loss of the resonance energies of the benzene rings.