EMAIL THIS PAGE TO A FRIEND
Radiation protection dosimetry
Preparation and emanation properties of an ion-exchanged solid thoron source.
PMID 22908351
Abstract
For the calibration purpose of thoron ((220)Rn) measurements, solid sources with constant (220)Rn emanation are generally desired. In this study, Th(4+) in the thorium nitrate powder was successfully exchanged to the surfaces of sodium benzenesulphonate resins with a bead diameter of ∼0.5 mm by using the ion-exchange method. Tests of (220)Rn emanating from the ion-exchanged resin indicated that the emanation rates were nearly constant, provided both the air humidity and air temperature were controlled. As the resin can be easily prepared to be of various sizes and activities of sources, the ion-exchanged resin is considered as a promising standard source for calibrations of (220)Rn measurements.