EMAIL THIS PAGE TO A FRIEND
Voprosy kurortologii, fizioterapii, i lechebnoi fizicheskoi kultury
[The influence of ultrasonic radiation on the factors of antimicrobial resistance in the women of reproductive age presenting with mycoplasma infection of the urogenital tract].
PMID 22908470
Abstract
The present clinical immunological study included 90 women of reproductive age presenting with mycoplasma infection of the lower urogenital tract. Low-frequency ultrasonic radiation was shown to successfully correct the disbalance in the system of congenital immunity that manifested itself as marked dysfunction of neutrophil granulocytes. The local application of low-frequency ultrasonic radiation normalized concentration of neutrophil defensins in cervical secretion as well as the number and function of neutrophils themselves.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59983
Josamycin, ≥90% (HPLC)
C42H69NO15
Y0000041
Josamycin, European Pharmacopoeia (EP) Reference Standard
C42H69NO15