EMAIL THIS PAGE TO A FRIEND
Natural product communications
Antioxidant compounds from Algerian Convolvulus tricolor (Convolvulaceae) seed husks.
PMID 22908568
Abstract
Seed husk extracts of Convolvulus tricolor L. (Convolvulaceae) afforded six compounds, identified for the first time from this plant: isorhamnetin 3-O-beta-D-galactopyranoside (1), isorhamnetin 3-O-beta-D-(6"-acetyl)-galactopyranoside (2), isorhamnetin 3-O-robinobioside (3), 3,4-di-O-caffeoylquinic acid (4), gentisic acid 5-O-glucoside (5), and scopoletin (6). Separation of compounds was carried out by CC and CPC. Structural elucidations were performed by HPLC-UV-DAD, HPLC-ESI/MS (negative mode) and NMR.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S2500
Scopoletin, ≥99%
C10H8O4
38332
Scopoletin, analytical standard
C10H8O4