EMAIL THIS PAGE TO A FRIEND
Organic letters
Total synthesis of pentosidine.
PMID 22909052
Abstract
Pentosidine, a biologically important advanced glycation endproduct, has been accessed in a rapid, high-yielding manner. The synthesis was accomplished via a six-step sequence starting with 3-amino-2-chloropyridine and features a palladium-catalyzed tandem cross-coupling/cyclization to construct the imidazo[4,5-b]pyridine core.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A93607
4-Azabenzimidazole, 99%
C6H5N3