EMAIL THIS PAGE TO A FRIEND
Allergy
Attenuated Bordetella pertussis BPZE1 protects against allergic airway inflammation and contact dermatitis in mouse models.
PMID 22909095
Abstract
We previously reported that prior nasal administration of highly attenuated Bordetella pertussis BPZE1 provides effective and sustained protection against lethal challenge with influenza A viruses. The protective effect was mediated by suppressing the production of major pro-inflammatory mediators. To further explore the anti-inflammatory properties of BPZE1, we investigated the effect of BPZE1 nasal pretreatment on two mouse models of allergic disease, allergic airway inflammation, and contact hypersensitivity (CHS). Allergic reactions were induced in mice nasally pretreated with live attenuated BPZE1 bacteria using the ovalbumin (OVA)-induced allergic airway inflammation and dinitrochlorobenzene (DNCB)-induced CHS models. Prior BPZE1 nasal treatment suppressed OVA-induced lung inflammation and inflammatory cell recruitment and significantly reduced IgE levels and cytokine production. Similarly, BPZE1 nasal pretreatment markedly inhibited ear swelling, skin inflammation, and production of pro-inflammatory cytokines in the DNCB-induced CHS model. For both models, we showed that BPZE1 pretreatment does not affect the sensitization phase. Upon challenge, BPZE1 pretreatment selectively reduced the level of cytokines whose production is increased and did not affect the basal level of other cytokines. Together, our observations suggest that BPZE1 pretreatment specifically targets those cytokine-producing effector cells that are recruited and involved in the inflammatory reaction. Our study demonstrates the broad anti-inflammatory properties of the attenuated B. pertussis BPZE1 vaccine candidate and supports its development as a promising agent to prevent and/or treat allergic diseases.