EMAIL THIS PAGE TO A FRIEND
Postgraduate medicine
Initial combination therapy with metformin plus colesevelam in drug-naïve Hispanic patients with early type 2 diabetes.
PMID 22913889
Abstract
To evaluate initial combination therapy with metformin plus colesevelam in drug-naïve Hispanic patients with type 2 diabetes mellitus. Patients self-identified as Hispanic from a previous study were included in this exploratory post hoc analysis. Patients were randomized to metformin plus colesevelam or metformin plus placebo. The primary efficacy parameter was the mean change in glycated hemoglobin (HbA1c) levels from baseline. Metformin plus colesevelam had a greater mean HbA1c reduction (-1.2 ± 0.1%) than metformin plus placebo (-0.8 ± 0.1%; P = 0.001) from mean baselines of 7.7% and 7.6%, respectively. Low-density lipoprotein cholesterol (LDL-C), non-high-density lipoprotein cholesterol, total cholesterol, and apolipoprotein (apo) B levels were also reduced (P < 0.0001 for all), while triglyceride (P < 0.0001) and apoA-I (P < 0.05) levels were increased with metformin plus colesevelam treatment compared with metformin plus placebo. With metformin plus colesevelam versus metformin plus placebo, more patients achieved an HbA1c of < 7.0% (75% vs 56%) and LDL-C of < 100 mg/dL (49% vs 14%; both P < 0.05). Metformin plus colesevelam may be an effective initial treatment option for Hispanic patients with early type 2 diabetes mellitus.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
241075
Allylamine, ≥99%
C3H7N
145831
Allylamine, 98%
C3H7N