EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Synthesis of a fluorescently tagged sialic acid analogue useful for live-cell imaging.
PMID 22914432
Abstract
A cytidine 5'-monophosphate (CMP)-sialic acid analogue carrying a fluorescent reporter group, an inhibitor of sialyltransferase, was synthesised in order to investigate glycan synthesis events in cells. The compound was found to be a substrate of a CMP-sialic acid transporter, and specific Golgi vesicles were visualised in the cells.