EMAIL THIS PAGE TO A FRIEND
Regional anesthesia and pain medicine
Duration and local toxicity of sciatic nerve blockade with coinjected site 1 sodium-channel blockers and quaternary lidocaine derivatives.
PMID 22914659
Abstract
Quaternary lidocaine derivatives (QLDs) have recently received much attention because of their potential application in prolonged or sensory-selective local anesthesia. However, associated tissue toxicity is an impeding factor that makes QLDs unfavorable for clinical use. Based on the proposed intracellular site of action, we hypothesized that nerve blocks obtained from lower concentrations of QLDs would be enhanced by the coapplication of extracellularly acting site 1 sodium-channel blocker, resulting in prolonged block duration but with minimal tissue toxicity. Quaternary lidocaine derivatives (QX-314 or QX-222), site 1 sodium-channel blockers (tetrodotoxin [30 μM] or saxitoxin [12.5 μM]), or both were injected in the vicinity of the sciatic nerve. Thermal nociceptive block was assessed using a modified hot plate test; motor block by a weight-bearing test. Tissue from the site of injection was harvested for histological assessment. Coapplication of 25 mM QX-314 or 100 mM QX-222 with site 1 sodium-channel blockers produced an 8- to 10- fold increase in the duration of nerve blocks (P < 0.05), compared with QLDs or site 1 blockers alone. Quaternary lidocaine derivatives elicited severe myotoxicity; this was not exacerbated by coinjection of the site 1 sodium-channel blockers. Coadministration of site 1 sodium-channel blockers and QLDs greatly prolongs the duration of peripheral nerve block without enhancing local tissue injury, but minimal myotoxicity still persists. It is not clear that the risks of QLDs are outweighed by the benefits in providing prolonged nerve blockade.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L5783
Lidocaine N-ethyl bromide, analytical standard, for drug analysis
C16H27BrN2O