EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
New excited state intramolecular proton transfer (ESIPT) dyes based on naphthalimide and observation of long-lived triplet excited states.
PMID 22914845
Abstract
A new excited state intramolecular proton transfer chromophore of naphthalimide (NI) conjugated 2-(2-hydroxyphenyl) benzothiazole (HBT) was prepared which shows red shifted absorption and long-lived triplet excited states.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
632589
2-(2-Hydroxyphenyl)benzothiazole, 97%
C13H9NOS