EMAIL THIS PAGE TO A FRIEND
Australian and New Zealand journal of medicine
Torsade de pointes associated with perhexiline maleate therapy.
PMID 2291733
Abstract
Multiform ventricular tachycardia (torsade de pointes) is a recognised proarrhythmic effect of drugs which prolong the QT interval. A case is now described for the first time where torsade de pointes occurred with the administration of the anti-anginal agent perhexiline maleate.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0120
Perhexiline maleate salt, ≥98% (HPLC)
C19H35N · C4H4O4