EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Synthesis of gold nanoparticles in niosomes.
PMID 22918054
Abstract
This work outlines a novel method for the synthesis of stable gold nanoparticles within the spatially confined region of vesicles. For the first time, Span/cholesterol based niosomes have been used for nanoparticle synthesis. The restricted geometry within niosomes prevents nanoparticle aggregation. The results have important implications for controlled delivery of nanoparticles for therapeutic applications.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6635
Span® 20