EMAIL THIS PAGE TO A FRIEND
Blood
ATRA plus arsenic gets another "A" in APL.
PMID 22918418
Abstract
In this issue of Blood, Iland et al report that the addition of arsenic trioxide during induction and consolidation can substantially reduce the amount of chemotherapy and the duration of consolidation to achieve excellent outcomes in patients with newly diagnosed acute promyelocytic leukemia (APL; see figure).