EMAIL THIS PAGE TO A FRIEND
Molecular Endocrinology
Dopamine synthesis and D3 receptor activation in pancreatic ?-cells regulates insulin secretion and intracellular [Ca(2+)] oscillations.
PMID 22918877