EMAIL THIS PAGE TO A FRIEND
Cardiovascular research
Upregulation of junctional adhesion molecule-A is a putative prognostic marker of hypertension.
PMID 22918977
Abstract
Establishing biochemical markers of pre-hypertension and early hypertension could help earlier diagnostics and therapeutic intervention. We assess dynamics of junctional adhesion molecule-A (JAM-A) expression in rat models of hypertension and test whether JAM-A expression could be driven by angiotensin (ANG) II and whether JAM-A contributes to the progression of hypertension. We also compare JAM-A expression in normo- and hypertensive humans. In pre-hypertensive and spontaneously hypertensive rats (SHRs), JAM-A protein was overexpressed in the brainstem microvasculature, lung, liver, kidney, spleen, and heart. JAM-A upregulation at early and late stages was even greater in the stroke-prone SHR. However, JAM-A was not upregulated in leucocytes and platelets of SHRs. In Goldblatt 2K-1C hypertensive rats, JAM-A expression was augmented before any increase in blood pressure, and similarly JAM-A upregulation preceded hypertension caused by peripheral and central ANG II infusions. In SHRs, ANG II type 1 (AT(1)) receptor antagonism reduced JAM-A expression, but the vasodilator hydralazine did not. Body-wide downregulation of JAM-A with Vivo-morpholinos in juvenile SHRs delayed the progression of hypertension. In the human saphenous vein, JAM-A mRNA was elevated in hypertensive patients with untreated hypertension compared with normotensive patients but reduced in patients treated with renin-angiotensin system antagonists. Body-wide upregulation of JAM-A in genetic and induced models of hypertension in the rat precedes the stable elevation of arterial pressure. JAM-A upregulation may be triggered by AT(1) receptor-mediated signalling. An association of JAM-A with hypertension and sensitivity to blockers of ANG II signalling were also evident in humans. We suggest a prognostic and possibly a pathogenic role of JAM-A in arterial hypertension.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H1753
Hydralazine hydrochloride
C8H8N4 · HCl