EMAIL THIS PAGE TO A FRIEND
Organic letters
Phorone A and isophorbasone A, sesterterpenoids isolated from the marine sponge Phorbas sp.
PMID 22920512
Abstract
A chemical investigation of a Korean marine sponge, Phorbas sp., yielded unprecedented sesterterpenoids phorone A (1) and isophorbasone A (2) along with ansellone B (3) and phorbasone A acetate (4). Their complete structures were elucidated by the combination of spectroscopic data and chemical manipulation. Phorone A (1) and isophorbasone A (2) have the new "phorane"(5) and "isophorbasane"(6) sesterterpenoid carbon skeletons, respectively. Ansellone B (3) and phorbasone A acetate (4) exhibited potent inhibitory activity on nitric oxide production in RAW 264.7 LPS-activated mouse macrophage cells with IC(50) values of 4.5 and 2.8 μM, respectively.