EMAIL THIS PAGE TO A FRIEND
Chemistry & biology
Targeted single-cell microchemical analysis: MS-based peptidomics of individual paraformaldehyde-fixed and immunolabeled neurons.
PMID 22921068
Abstract
Pinpointing a specific cell from within a relatively uniform cell population to determine its chemical content presents a challenging bioanalytical task. Immunocytochemistry is the classical method used to localize specific molecules and, hence, selected cells. Mass spectrometry also probes endogenous molecules such as neuropeptides within a cell. Here, these two approaches are hyphenated to allow microchemical analysis of immunocytochemical-selected peptidergic neurons. This two-step strategy utilizes antibody-based localization of cells containing selected biomarkers to isolate the cell(s) of interest, followed by peptidomic analysis via mass spectrometry. Applicable to a broad range of analyte and cell types, the strategy was used to successfully profile neuropeptides from individual immunostained insect neurons stored for up to 2 weeks as well as from tissues preserved for 42 weeks.