EMAIL THIS PAGE TO A FRIEND
Journal of microbiological methods
Development of a novel primer combination to detect pathogenic Leptospira by loop-mediated isothermal amplification.
PMID 22922651
Abstract
Loop-mediated isothermal amplification (LAMP) has been used for the detection of leptospiral 16S rDNA. Here we report the development of the LAMP method for leptospiral 16S rDNA detection which can be interpreted easily within 90 min by visualizing the fluorescence of calcein. The lower limit of detection (LLOD) is 10-100 copies.