EMAIL THIS PAGE TO A FRIEND
Journal of neurochemistry
Molecular determinants of A2AR-D2R allosterism: role of the intracellular loop 3 of the D2R.
PMID 22924752
Abstract
In the CNS, an antagonistic interaction has been shown between adenosine A(2A) and dopamine D(2) receptors (A(2A)Rs and D(2)Rs) that may be relevant both in normal and pathological conditions (i.e., Parkinson's disease). Thus, the molecular determinants mediating this receptor-receptor interaction have recently been explored, as the fine tuning of this target (namely the A(2A)R/D(2)R oligomer) could possibly improve the treatment of certain CNS diseases. Here, we used a fluorescence resonance energy transfer-based approach to examine the allosteric modulation of the D(2)R within the A(2A)R/D(2)R oligomer and the dependence of this receptor-receptor interaction on two regions rich in positive charges on intracellular loop 3 of the D(2)R. Interestingly, we observed a negative allosteric effect of the D(2)R agonist quinpirole on A(2A)R ligand binding and activation. However, these allosteric effects were abolished upon mutation of specific arginine residues (217-222 and 267-269) on intracellular loop 3 of the D(2)R, thus demonstrating a major role of these positively charged residues in mediating the observed receptor-receptor interaction. Overall, these results provide structural insights to better understand the functioning of the A(2A)R/D(2)R oligomer in living cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q102
(−)-Quinpirole hydrochloride, ≥98% (HPLC), solid
C13H21N3 · HCl