EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
New fluorescent reagents specific for Ca(2+)-binding proteins.
PMID 22925895
Abstract
Ca(2+) carries information pivotal to cell life and death via its interactions with specific binding sites in a protein. We previously developed a novel photoreactive reagent, azido ruthenium (AzRu), which strongly inhibits Ca(2+)-dependent activities. Here, we synthesized new fluorescent ruthenium-based reagents containing FITC or EITC, FITC-Ru and EITC-Ru. These reagents were purified, characterized and found to specifically interact with and markedly inhibit Ca(2+)-dependent activities but not the activity of Ca(2+)-independent reactions. In contrast to many reagents that serve as probes for Ca(2+), FITC-Ru and EITC-Ru are the first fluorescent divalent cation analogs to be synthesized and characterized that specifically bind to Ca(2+)-binding proteins and inhibit their activity. Such reagents will assist in characterizing Ca(2+)-binding proteins, thereby facilitating better understanding of the function of Ca(2+) as a key bio-regulator.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45245
Eosin 5-isothiocyanate, ≥95.0% (UV)
C21H7Br4NO5S