EMAIL THIS PAGE TO A FRIEND
Annales francaises d'anesthesie et de reanimation
[Iode poisoning after several bandages].
PMID 22925938
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
109452
Iodoform, 99%
CHI3