EMAIL THIS PAGE TO A FRIEND
Journal of clinical psychopharmacology
Mania related to mesterolone in a previously mentally healthy person.
PMID 22926621
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M7655
Mesterolone, analytical standard
C20H32O2