EMAIL THIS PAGE TO A FRIEND
Evidence-based complementary and alternative medicine : eCAM
The chinese herbal decoction danggui buxue tang inhibits angiogenesis in a rat model of liver fibrosis.
PMID 22927877
Abstract
In this study, we investigated the anti-angiogenic effect of the Chinese herbal decoction Danggui Buxue Tang (DBT; Radix Astragali and Radix Angelicae sinensis in 5 : 1 ratio) in a rat model of liver fibrosis, in order to elucidate its mechanisms of action against liver fibrosis. Liver fibrosis was induced with CCl(4) and high-fat food for 6 weeks, and the rats were treated with oral doses of DBT (6 g raw herbs/kg/d) and N-Acetyl-L-cysteine (NAC; 0.1 g/kg/d). The results showed that both DBT and NAC attenuated liver fibrosis and neo-angiogenesis. Furthermore, DBT and NAC improved SOD activity but decreased MDA content and 8-OH-dG in fibrotic livers, with DBT being more effective than NAC. DBT decreased the expression of VEGF, Ang1 and TGF-β1 and their signaling mediators, whereas NAC had no effect on VEGF and VEGFR2 expression. Both DBT and NAC reduced HIF-1α gene and protein expression in fibrotic livers, with DBT being more effective. These data clearly demonstrate that the anti-fibrotic properties of DBT are related to its ability to inhibit angiogenesis and its anti-angiogenic mechanisms are associated with improving oxidative stress, regulating the expression and signaling of angiogenic factors, and especially modulating HIF-1α in fibrotic livers.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H5653
8-Hydroxy-2′-deoxyguanosine, ≥98% (TLC)
C10H13N5O5