EMAIL THIS PAGE TO A FRIEND
IARC monographs on the evaluation of carcinogenic risks to humans
Trichlormethine (trimustine hydrochloride).
PMID 2292796
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T85308
Tris(2-chloroethyl)amine hydrochloride, 98%
C6H12Cl3N · HCl