EMAIL THIS PAGE TO A FRIEND
Diabetes technology & therapeutics
Predicting outcomes and assessing control with alternate glycemic markers.
PMID 22928613
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A7165
1,5-Anhydro-D-sorbitol, crystalline
C6H12O5