EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Lewis acid trapping of an elusive copper-tosylnitrene intermediate using scandium triflate.
PMID 22928636
Abstract
High-valent copper-nitrene intermediates have long been proposed to play a role in copper-catalyzed aziridination and amination reactions. However, such intermediates have eluded detection for decades, preventing the unambiguous assignments of mechanisms. Moreover, the electronic structure of the proposed copper-nitrene intermediates has also been controversially discussed in the literature. These mechanistic questions and controversy have provided tremendous motivation to probe the accessibility and reactivity of Cu(III)-NR/Cu(II)N(•)R species. In this paper, we report a breakthrough in this field that was achieved by trapping a transient copper-tosylnitrene species, 3-Sc, in the presence of scandium triflate. The sufficient stability of 3-Sc at -90 °C enabled its characterization with optical, resonance Raman, NMR, and X-ray absorption near-edge spectroscopies, which helped to establish its electronic structure as Cu(II)N(•)Ts (Ts = tosyl group) and not Cu(III)NTs. 3-Sc can initiate tosylamination of cyclohexane, thereby suggesting Cu(II)N(•)Ts cores as viable reactants in oxidation catalysis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261262
Scandium, powder, 99.9% trace rare earth metals basis
Sc