EMAIL THIS PAGE TO A FRIEND
Proteomics
Interferometric silicon biochips for label and label-free DNA and protein microarrays.
PMID 22930463
Abstract
Protein and DNA microarrays hold the promise to revolutionize the field of molecular diagnostics. Traditional microarray applications employ labeled detection strategies based on the use of fluorescent and chemiluminescent secondary antibodies. However, the development of high throughput, sensitive, label-free detection techniques is attracting attention as they do not require labeled reactants and provide quantitative information on binding kinetics. In this article, we will provide an overview of the recent author's work in label and label-free sensing platforms employing silicon/silicon oxide (Si/SiO(2)) substrates for interferometric and/or fluorescence detection of microarrays. The review will focus on applications of Si/SiO(2) with controlled oxide layers to (i) enhance the fluorescence intensity by optical interferences, (ii) quantify with sub-nanometer accuracy the axial locations of fluorophore-labeled probes tethered to the surface, and (iii) detect protein-protein interactions label free. Different methods of biofunctionalization of the sensing surface will be discussed. In particular, organosilanization reactions for monodimensional coatings and polymeric coatings will be extensively reviewed. Finally, the importance of calibration of protein microarrays through the dual use of labeled and label-free detection schemes on the same chip will be illustrated.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF72027266
Silicon, disks, 13mm, thickness 0.38mm, single crystal, 100%
Si
GF16286227
Silicon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%
Si
GF46268433
Silicon, disks, 15.9mm, thickness 0.38mm, single crystal, n-type, 100%
Si
GF37993595
Silicon, rod, 100mm, diameter 10.0mm, single crystal - random orientation, 100%
Si
GF84708681
Silicon, rod, 100mm, diameter 12.7mm, single crystal - random orientation, 100%
Si
GF64686998
Silicon, rod, 100mm, diameter 2.0mm, crystalline, 100%
Si
GF04814951
Silicon, rod, 100mm, diameter 25mm, crystalline, 100%
Si
GF22929307
Silicon, rod, 100mm, diameter 5.0mm, crystalline, 100%
Si
GF16363391
Silicon, rod, 10mm, diameter 2.0mm, crystalline, 100%
Si
GF66465276
Silicon, rod, 25mm, diameter 2.0mm, crystalline, 100%
Si
GF43555664
Silicon, rod, 25mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF70438518
Silicon, rod, 40mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF05482852
Silicon, rod, 50mm, diameter 2.0mm, crystalline, 100%
Si
GF91515034
Silicon, rod, 50mm, diameter 3.15mm, single crystal - random orientation, 100%
Si
GF14076930
Silicon, rod, 50mm, diameter 5.0mm, crystalline, 100%
Si
GF28448691
Silicon, rod, 50mm, diameter 5mm, single crystal, -100, 99.999%
Si
GF40945754
Silicon, rod, 50mm, diameter 5mm, single crystal, -111, 99.999%
Si
GF64342188
Silicon, rod, 50mm, diameter 6mm, single crystal, -111, 99.999%
Si
GF10907377
Silicon, rod, 80mm, diameter 20mm, single crystal, -100, 99.999%
Si
GF91141792
Silicon, sheet, 10x10mm, thickness 0.5mm, single crystal, -100, 100%
Si
GF71817543
Silicon, sheet, 10x10mm, thickness 0.6mm, single crystal, -100, 100%
Si
GF75683212
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -100, 100%
Si
GF84840004
Silicon, sheet, 14x14mm, thickness 1.0mm, single crystal, -111, 100%
Si
GF20311254
Silicon, sheet, 25x25mm, thickness 1.0mm, polycrystalline, 99.999%
Si
GF41762493
Silicon, sheet, 25x25mm, thickness 1.0mm, single crystal, -100, 100%
Si
GF85621923
Silicon, sheet, 40x40mm, thickness 3.0mm, single crystal, p-type, 100%
Si
GF73529051
Silicon, sheet, 52x52mm, thickness 1.0mm, polycrystalline, 99.999%
Si
GF23955622
Silicon, single crystal, -100, n-type, phosphorus doped, diameter 150mm, length 0.65mm, 100%
Si
GF06693756
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.50mm, 100%
Si
GF68832805
Silicon, single crystal, -111, n-type, phosphorus doped, diameter 76.2mm, length 0.75mm, 100%
Si