EMAIL THIS PAGE TO A FRIEND
Cytometry. Part A : the journal of the International Society for Analytical Cytology
Multicolor flow cytometry analysis of the proliferations of T-lymphocyte subsets in vitro by EdU incorporation.
PMID 22930591
Abstract
EdU (5-ethynyl-2'-deoxyuridine) incorporation has proved advantageous in the studies of cell kinetics, DNA synthesis, and cellular proliferation in vitro and in vivo compared to [(3) H]thymidine incorporation and BrdU (5-bromo-2'-deoxyuridine) incorporation. Here, we describe a method that combines EdU incorporation and immunostaining with flow cytometric analysis to detect the proliferations of T lymphocyte subsets in vitro and optimized the assay's conditions. We found that the number of EdU(+) cells were associated with EdU concentration, incubation time, and the volume of Click reaction solution, the best EdU concentration 10-50 μM, the optimal incubation time 8-12 h and the proper volume of Click volume 100 μl for labeling 1 × 10(6) lymphocytes. Fixation was better to be performed before permeabilization, not together with. Furthermore, the permeabilization detergent reagent, PBS with 0.05% saponin was better than Tris buffer saline (TBS) with 0.1% Triton X-100. In addition, sufficient wash with PBS with 0.05% saponin has no influence on the staining of EdU(+) cells. Also, the lymphocytes incorporating EdU could be stored at 4°C, -80°C, and in liquid nitrogen up to 21 days. The present study will aid in optimization of flow cytometry assay to detect the proliferations of T cell subsets by EdU incorporation and the labeling of cell surface antigens.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5412
2′-Deoxyuridine, ≥98.5%
C9H12N2O5