EMAIL THIS PAGE TO A FRIEND
Clinical pediatrics
A Harry Potter fan.
PMID 22930779
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P1000000
Phenoxymethylpenicillin, European Pharmacopoeia (EP) Reference Standard
C16H18N2O5S