EMAIL THIS PAGE TO A FRIEND
Zhonghua lao dong wei sheng zhi ye bing za zhi = Zhonghua laodong weisheng zhiyebing zazhi = Chinese journal of industrial hygiene and occupational diseases
[Studing the health status of workers occupationally exposed to carbon disulfide].
PMID 22931773
Abstract
To study the effects of carbon disulfide (CS(2)) on the health of workers, and to provide the basis for the further preventive measures. The occupational health examination of 881 workers exposed to CS(2) in a chemical fiber factory was carried out according to the national technical standard of occupational health surveillance. The time weighted average concentrations (TWA) of CS(2) in the workshops of short silk and long silk exceeded the national standard, The workers in these two workshops served as the high exposure group. The workers of the other workshops were classified as low exposure group, in which TWA met the national standard. All exposed workers were also divided into long-term exposure group and short-term exposure group on the basis of average exposure years (16 years). The statistical analysis was used by t test, χ(2), and fisher exact test. The symptom rates of numbness, fatigue, dizziness, insomnia and headache were 12.5%, 8.5%, 8.2%, 7.5%, and 7.2% respectively. The abnormal rates of superficial sensation, three fibrillation, achilles tendon reflex, patellar reflex, EMG, hypertension, triglycerides, low-density lipoprotein, and apolipoprotein B were 33.0%, 26.1%, 20.8%, 18.6%, 10.8%, 33.4%, 24.5%, 17.0% and 9.3% respectively. Among the high exposure group, the abnormal detectable rates of dizziness, headache, fatigue, insomnia, numbness, achilles tendon reflex, superficial and deep sensation, EMG, hypertension, ECG, total cholesterol, triglyceride and low density lipoprotein in long-term exposure group were significantly higher than those in short-term exposure group (P < 0.05 or P < 0.01). However, among the low exposure group, the abnormal rates of the insomnia achilles tendon reflex and superficial sensation in long term exposure group were significantly higher than those of short term exposure group (P < 0.05 or P < 0.01). The detected rates of hypertension, triglycerides, low-density lipoprotein, creatinine and uric acid were in males significantly greater than those in females (P < 0.05 or P < 0.01). Long-term exposure to high concentrations of CS(2) could lead to the damage of nervous system, elevate blood pressure and promote the development and progression of hyperlipemia and atherosclerosis. Furthermore, CS(2) had greater effects on blood pressure and lipid in males than in females.