EMAIL THIS PAGE TO A FRIEND
Radiation oncology (London, England)
MVP expression in the prediction of clinical outcome of locally advanced oral squamous cell carcinoma patients treated with radiotherapy.
PMID 22931894
Abstract
To explore the role of Major Vault Protein (MVP) in oral cavity squamous cell carcinoma patients. 131 consecutive patients suffering from oral cavity squamous cell carcinoma were included in the study. In the whole series, the mean follow-up for survivors was 123.11 ± 40.36 months. Patients in tumour stages I and II were referred to surgery; patients in stage III-IV to postoperative radiotherapy (mean dose = 62.13 ± 7.74 Gy in 1.8-2 Gy/fraction). MVP expression was studied by immunohistochemistry in paraffin-embedded tumour tissue. MVP expression was positive in 112 patients (85.5%) and no relation was found with clinic pathological variables. MVP overexpression (those tumours with moderate or strong expression of the protein) was related to insulin-like growth factor receptor-1 (IGF-1R) expression (P = 0.014). Tumour stage of the disease was the most important prognostic factor related to survival. Tumours overexpressing MVP and IGF-1R were strongly related to poor disease-free survival (P = 0.008, Exp(B) = 2.730, CI95% (1.302-5.724)) and cause-specific survival (P = 0.014, Exp(B) = 2.570, CI95% (1.215-5.437)) in patients achieving tumour stages III-IV, in multivariate analysis. MVP and IGF-1R expression were related in oral squamous cell carcinoma and conferred reduced long-term survival in patients suffering from advanced stages of the disease.