Molecular cancer therapeutics
Perillyl alcohol for the treatment of temozolomide-resistant gliomas.
PMID 22933703
Perillyl alcohol (POH) is a monoterpene that has been used orally for the treatment of systemic cancer. However, when used orally significant gastrointestinal side effects and lack of overall efficacy were documented. Recently, in a phase II trial in Brazil for the treatment of temozolomide (TMZ)-resistant malignant gliomas, POH was well tolerated when administered intranasally. The present study explores the effects and mechanisms of POH on TMZ-sensitive and TMZ-resistant glioma cells. In vitro studies showed that POH was cytotoxic to TMZ-resistant as well as TMZ-sensitive glioma cells, and this effect was independent of O(6)-methylguanine-DNA methyltransferase expression. POH induced cytotoxicity, in part, through the endoplasmic reticulum (ER) stress pathway as shown by the increased expression of glucose-regulated protein-78 (GRP78), activating transcription factor 3, and C/EBP-homologous protein. In addition, POH impeded survival pathways, such as mTOR and Ras. As well, POH reduced the invasive capacity of sensitive and resistant glioma cells. POH alone and/or in combination with other ER stress-inducing cytotoxic drugs (i.e., 2, 5-dimethyl-celecoxib, nelfinavir) further induced apoptosis in TMZ-sensitive and TMZ-resistant glioma cells. To show whether intranasal delivery of POH was effective for the treatment of TMZ-resistant gliomas, animals bearing intracranial tumors were given POH intranasally. Animals treated through intranasal administration of POH exhibited a decrease in tumor growth and an increase in survival. Our data show that POH is an effective anti-glioma cytotoxic agent for TMZ-resistant gliomas when administered intranasally.