EMAIL THIS PAGE TO A FRIEND
Retina (Philadelphia, Pa.)
Characteristics of retained subretinal perfluoro-n-octane on optical coherence tomography.
PMID 22936004
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
359238
Perfluorooctane, 98%
C8F18