EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Convergent synthesis of conjugated 1,2-disubstituted E-allylic alcohols from two aldehydes and methylenetriphenylphosphorane.
PMID 22936294
Abstract
β-Lithiooxyphosphonium ylides, made in situ from an aldehyde and methylenetriphenylphosphorane, react with a second aldehyde to form E-allylic alcohols. α-Branching and α,β-unsaturation in the second aldehyde, together with the lack of further substitution on the phosphorane carbon play important roles in selectivity. A range of these aldehydes, in addition to aromatic aldehydes as the second aldehyde also provided synthetically useful access to E-allylic alcohols.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
459798
Allyl alcohol, ≥98.5%
C3H6O
442445-U
Allyl alcohol, analytical standard
C3H6O