EMAIL THIS PAGE TO A FRIEND
Journal of food science
The effectiveness of antibrowning dip treatments to reduce after-cooking darkening in potatoes.
PMID 22938507
Abstract
After-cooking darkening (ACD) is an inherent and undesirable trait that develops in cooked potatoes. The objective of this study was to evaluate the effectiveness of sodium acid sulfate (SAS) dip treatments compared to other antigraying treatments and a control to reduce ACD in boiled, Katahdin potatoes. Dip treatments were applied for 3 min prior to boiling and included: 3% SAS, 3% citric acid (CA), 3% sodium acid pyrophosphate (SAPP), along with a distilled water control. SAS- and CA-treated potatoes had slightly, but significantly (P ≤ 0.05) higher b* and chroma values, which indicates a more intense yellow potato color, with less graying, compared to the control. SAS- and CA-treated potatoes also had significantly (P≤ 0.001) lower pH values for inner and outer potato surfaces than the control. No significant (P > 0.05) differences were detected for total phenolic or mineral contents among treatments. CA and SAPP samples had slightly, but significantly (P≤ 0.05) higher moisture contents than the control. Sensory test results showed no significant differences for color, aftertaste, or overall acceptability. However, CA-treated samples were rated significantly (P≤ 0.05) lower for flavor than all other treatments and panelists commented on sour notes. CA- and SAS-treated potatoes were scored slightly, but significantly lower for texture than other treatments due to a waxy outer layer. However, SAS was the most acidic dip treatment, but did not significantly affect flavor. Overall, results suggest that SAS was similarly accepted by consumers in comparison to CA and SAPP, which is the industry standard to reduce ACD. After-cooking darkening (ACD) is an undesirable potato trait that occurs after potatoes have been processed. Sodium acid pyrophosphate (SAPP) has been used as the industry standard to reduce ACD. Sodium acid sulfate (SAS) treatments prior to boiling appeared to be comparable to SAPP and citric acid in effectiveness to reduce ACD. SAS did not negatively affect the flavor of boiled potato samples according to sensory results. The SAS treatment may be more beneficial for potatoes intended for potato salad products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
71657
Sodium bisulfate monohydrate, puriss. p.a., ≥99.0% (T)
HNaO4S · H2O
233714
Sodium bisulfate monohydrate, ReagentPlus®, 99%
HNaO4S · H2O
221368
Sodium pyrophosphate decahydrate, ACS reagent, ≥99%
Na4O7P2 · 10H2O
P8135
Sodium pyrophosphate dibasic, practical grade
H2Na2O7P2
71501
Sodium pyrophosphate dibasic, BioUltra, ≥99.0% (T)
H2Na2O7P2
S6422
Sodium pyrophosphate tetrabasic decahydrate, BioXtra, 99.0-103.0%
Na4O7P2 · 10H2O
71515
Sodium pyrophosphate tetrabasic decahydrate, BioUltra, ≥99.5% (T)
Na4O7P2 · 10H2O
30411
Sodium pyrophosphate tetrabasic decahydrate, puriss. p.a., ≥99%
Na4O7P2 · 10H2O
239313
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Na2O4S
238597
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Na2O4S
S9627
Sodium sulfate, ReagentPlus®, ≥99.0%
Na2O4S
31481
Sodium sulfate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.0%
Na2O4S
13464
Sodium sulfate, puriss., meets analytical specification of Ph. Eur., BP, USP, anhydrous, 99.0-100.5% (calc. to the dried substance)
Na2O4S
403008
Sodium sulfate decahydrate, ACS reagent, ≥99.0%
Na2O4S · 10H2O
246980
Sodium sulfate decahydrate, reagent grade, 97%
Na2O4S · 10H2O
71969
Sodium sulfate decahydrate, BioUltra, ≥99.0% (calc. on dry substance, T)
Na2O4S · 10H2O
71970
Sodium sulfate decahydrate, puriss. p.a., crystallized, ≥99.0% (calc. based on dry substance, T)
Na2O4S · 10H2O
13571
Sodium sulfate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.0% (calc. to the dried substance)
Na2O4S · 10H2O