EMAIL THIS PAGE TO A FRIEND
Analytica chimica acta
Detoxification of organophosphate residues using phosphotriesterase and their evaluation using flow based biosensor.
PMID 22938607
Abstract
Among known pesticide groups, organophosphates (OPs) have grasped attention due to their hazardous nature and their applications as pesticides and chemical weapons. This work presents the development of cost-effective column based biosensor for detoxification of OPs in water and milk. Enzyme phosphotriesterase (PTE) was immobilized on an activated Sepharose 4B via covalent coupling using an Omnifit glass column. Three different OPs, ethyl paraoxon (EPOx), malaoxon (MAO) and chlorpyriphos-oxon (CPO) were spiked in water and milk to test the detoxification of OPs. Mixtures of these pesticides were also tested to check the cumulative detoxification in the real samples. The efficiency of detoxification was evaluated using a highly sensitive acetylcholinesterase (AChE) B394 biosensor based flow system. The column conditions were optimized for the detoxification studied. The method was shown to be promising when we tested real milk samples spiked with OPs. Detoxification obtained in milk was up to 86% whereas in water, 100% detoxification was obtained.