EMAIL THIS PAGE TO A FRIEND
European journal of internal medicine
Antiplatelet treatment in primary and secondary stroke prevention in women.
PMID 22939800
Abstract
Stroke is a leading cause of death worldwide and the first cause of disability in the Western world. Over the last 20 years, antiplatelet agents have reduced overall stroke rates in primary and secondary prevention in men. However, this has not been the case for women. In this narrative review, the most widely used antiplatelet therapies for primary and secondary prevention in stroke, excluding cardioembolic stroke, will be outlined. First, the largest randomised controlled trials will be analysed as well as the enrolment percentages of women. Second, analyses on sex-interaction effects in each study will be examined. Moreover, the Authors will discuss the need to develop targeted antiplatelet therapies specifically for women. Based on current results, the most randomised clinical trials and meta-analyses on antiplatelet agents in cerebrovascular disease have not performed sub-analyses on sex-related differences and this is mainly because women were underrepresented. Despite this, antiplatelet agents are considered to be equally effective for both sexes in primary and secondary stroke prevention. Finally, aspirin is the most widely studied antiplatelet in women and has been shown to provide greater benefit for women as primary prevention of ischemic stroke without a significant increased risk in haemorrhage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6580 Triflusal, ≥98% (HPLC), powder
C10H7F3O4