EMAIL THIS PAGE TO A FRIEND
Optics letters
Double-layered nanoparticle stacks for spectro-electrochemical applications.
PMID 22940963
Abstract
Here we present a surface based on double-layered nanoparticle stacks suitable for spectro-electrochemical applications. The structure is formed on a continuous gold layer by a two-dimensional periodic array of stacks of gold and tantalum pentoxide nanodisks. Reflection spectra in the visible wavelength region showed the multiple-resonant nature of surface plasmon (SP) excitations in the nanostructure, which is in good agreement with simulations based on a finite-difference-time-domain method. The multiple SP resonances can be tuned to various wavelength regions, which are required for simultaneous enhancement at excitation and emission wavelengths. Cyclic voltammetry measurements on the nanostructure proved the applicability of electrochemical methods involving interfacial redox processes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
702587
Potassium ferricyanide(III), powder or chunks, <10 μm, 99%
455946
Potassium hexacyanoferrate(III), 99.98% trace metals basis
C6FeK3N6
244023
Potassium hexacyanoferrate(III), ACS reagent, ≥99.0%
C6FeK3N6
60299
Potassium hexacyanoferrate(III), BioUltra, ≥99.0% (RT)
C6FeK3N6
31253
Potassium hexacyanoferrate(III), puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99%
C6FeK3N6
P8131
Potassium hexacyanoferrate(III), ReagentPlus®, ~99%
C6FeK3N6
262889
Tantalum, foil, thickness 1.0 mm, ≥99.9% trace metals basis
Ta
357251
Tantalum, foil, thickness 0.5 mm, ≥99.9% trace metals basis
Ta
262897
Tantalum, foil, thickness 0.25 mm, ≥99.9% trace metals basis
Ta
357243
Tantalum, foil, thickness 0.05 mm, ≥99.9% trace metals basis
Ta
262919
Tantalum, foil, thickness 0.025 mm, ≥99.9% trace metals basis
Ta
545007
Tantalum, powder, 99.99% trace metals basis
Ta
692824
Tantalum, powder, 60-100 mesh, 99.9%
Ta
262846
Tantalum, powder, −325 mesh, 99.9% trace metals basis
Ta
593486
Tantalum, nanopowder, <25 nm particle size (BET), ≥99% trace metals basis
Ta
262854
Tantalum, rod, diam. 6.35 mm, ≥99.9% trace metals basis
Ta
356980
Tantalum, rod, diam. 3.2 mm, ≥99.9% trace metals basis
Ta
262862
Tantalum, wire, diam. 1.0 mm, ≥99.9% trace metals basis
Ta
357006
Tantalum, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Ta