EMAIL THIS PAGE TO A FRIEND
International journal of systematic and evolutionary microbiology
Roseovarius sediminilitoris sp. nov., isolated from seashore sediment.
PMID 22941298
Abstract
A Gram-negative, motile and ovoid- to rod-shaped bacterial strain, designated M-M10(T), was isolated from a seashore sediment collected from the South Sea, South Korea. Strain M-M10(T) grew optimally at pH 7.0-8.0, at 30 °C and in the presence of 2 % (w/v) NaCl. The phylogenetic trees based on 16S rRNA gene sequences revealed that strain M-M10(T) clustered with the type strains of Roseovarius crassostreae, Roseovarius halocynthiae and Roseovarius marinus, with which it exhibited sequence similarities of 97.4, 97.3 and 95.1 %, respectively. It exhibited 93.2-95.1 % sequence similarity to the type strains of the other species of the genus Roseovarius. Strain M-M10(T) contained Q-10 as the predominant ubiquinone and C18 : 1ω7c and C16 : 0 as the major fatty acids, as observed in the genus Roseovarius. The polar lipid profile of strain M-M10(T) was similar to that of Roseovarius tolerans DSM 11457(T). The DNA G+C content of strain M-M10(T) was 63.0 mol% and its mean DNA-DNA relatedness values with Roseovarius crassostreae DSM 16950(T) and Roseovarius halocynthiae MA1-10(T) were 16 % and 22 %, respectively. Differential phenotypic properties, together with the phylogenetic and genetic distinctiveness, demonstrated that strain M-M10(T) is distinct from other species of the genus Roseovarius. On the basis of the data presented, strain M-M10(T) is considered to represent a novel species of the genus Roseovarius, for which the name Roseovarius sediminilitoris sp. nov. is proposed. The type strain is M-M10(T) ( = KCTC 23959(T) = CCUG 62413(T)).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C8081
Coenzyme Q2, ≥90%
C19H26O4