EMAIL THIS PAGE TO A FRIEND
Nanoscale
Click chemistry: a new facile and efficient strategy for the preparation of Fe3O4 nanoparticles covalently functionalized with IDA-Cu and their application in the depletion of abundant protein in blood samples.
PMID 22941423
Abstract
In this study, we report a novel method to synthesize core-shell structured Fe(3)O(4) nanoparticles (NPs) covalently functionalized with iminodiacetic acid (IDA) via click chemistry between the azide and alkyne groups and charged with Cu(2+). Firstly, the Fe(3)O(4)@SiO(2) NPs were obtained using tetraethoxysilane (TEOS) to form a silica shell on the surface of the Fe(3)O(4) core. The azide group-modified Fe(3)O(4)@SiO(2) NPs were obtained by a sol-gel process using 3-azidopropyltriethoxysilane (AzPTES) as the silane agent. Fe(3)O(4)@SiO(2)-N(3) was directly reacted with N-propargyl iminodiacetic via click chemistry, in the presence of a Cu(I) catalyst, to acquire the IDA-modified Fe(3)O(4) NPs. Finally, through the addition of Cu(2+), the Fe(3)O(4)@SiO(2)-IDA-Cu NP product was obtained. The morphology, structure and composition of the NPs were characterized by transmission electron microscopy (TEM), X-ray powder diffraction (XRD), Fourier transform infrared (FTIR) spectroscopy and X-ray photoelectron spectroscopy (XPS). The resulting NPs showed a strong magnetic response to an externally applied magnetic field, a high adsorption capacity and excellent specificity towards hemoglobin (Hb). In addition, the Fe(3)O(4)@SiO(2)-IDA-Cu NPs can be used for the selective removal of abundant Hb protein in bovine and human blood samples.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
56781
Iminodiacetic acid, purum, ≥98.0% (T)
C4H7NO4
220000
Iminodiacetic acid, 98%
C4H7NO4