EMAIL THIS PAGE TO A FRIEND
Archives of pharmacal research
Design and synthesis of new 8-anilide theophylline derivatives as bronchodilators and antibacterial agents.
PMID 22941478
Abstract
Theophylline derivatives have long been recognized as potent bronchodilators for the relief of acute asthma. Recently, it was found that bacterial infection has a role in asthma pathogenesis. The present work involves the design and synthesis of 8-substituted theophylline derivatives as bronchodilators and antibacterial agents. The chemical structures of these compounds were elucidated by IR, (1)H-NMR, mass spectrometry, and elemental analyses. The bronchodilator activity was evaluated using acetylcholine-induced bronchospasm in guinea pigs, and most of the compounds showed significant anti-bronchoconstrictive activity in comparison with standard aminophylline. In addition, the antibacterial activity of all the target compounds was investigated in vitro against Gram-positive and Gram-negative bacteria using ampicillin as a reference drug. Results showed that some of the tested compounds possessed significant antibacterial activity. A pharmacophore model was computed to obtain useful insight into the essential structural features of bronchodilator activity. A structure activity relationship was also discussed.