EMAIL THIS PAGE TO A FRIEND
FEMS immunology and medical microbiology
Nonencapsulated Streptococcus pneumoniae resists extracellular human neutrophil elastase- and cathepsin G-mediated killing.
PMID 22943431
Abstract
Although the Streptococcus pneumoniae polysaccharide capsule is an important virulence factor, ~ 15% of carriage isolates are nonencapsulated. Nonencapsulated S. pneumoniae are a cause of mucosal infections. Recent studies have shown that neutrophils kill S. pneumoniae predominately through neutrophil proteases, such as elastase and cathepsin G. Another recent finding is that nonencapsulated pneumococci have greater resistance to resist cationic antimicrobial peptides that are important in mucosal immunity. We here show that nonencapsulated pneumococci have greater resistance to extracellular human neutrophil elastase- and cathepsin G-mediated killing than isogenic encapsulated pneumococci. Resistance to extracellular neutrophil protease-mediated killing is likely to be of greater relative importance on mucosal surfaces compared to other body sites.