EMAIL THIS PAGE TO A FRIEND
Carbohydrate polymers
Studies on the characteristic and activity of low-molecular fragments from zymosan.
PMID 22944396
Abstract
Zymosan was hydrolysed with HCl and fractionated by ultrafiltration and dialysis to obtain water-soluble fragments A, B and C. Physical and chemical analyses showed that these fractions are composed primarily of glucose and have molecular weights of 8 kDa, 5 kDa and 2 kDa, respectively. A glycosidic linkage analysis indicated that they are mainly composed of β-1,3-glucans. Fragment A, which has the highest molecular weight, contains approximately 30% β-1,6-linked glucans, but fragment C is almost entirely composed of linear β-1,3-glucan chains. The anti-chronic atrophic gastritis activity experiments showed that fragment A has significant activity, the activity of zymosan is quite low and the activities of fragments B and C are in between those of fragment A and zymosan.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89862
β-1,3-Glucan from Euglena gracilis