EMAIL THIS PAGE TO A FRIEND
Chemistry (Weinheim an der Bergstrasse, Germany)
A general palladium-catalyzed carbonylative synthesis of 2-alkylbenzoxazinones from 2-bromoanilines and acid anhydrides.
PMID 22945892
Abstract
(C), its (O)K! An efficient palladium-catalyzed carbonylative synthesis of 2-alkylbenzoxazinones has been developed. By starting from 2-bromoanilines and acid anhydrides, the corresponding products were isolated in good yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
180025
3-Bromoaniline, 98%
C6H6BrN