EMAIL THIS PAGE TO A FRIEND
Journal of zoo and wildlife medicine : official publication of the American Association of Zoo Veterinarians
Tolazoline-induced apnea in mule deer (Odocoileus hemionus).
PMID 22946377
Abstract
Eighteen mule deer (Odocoileus hemionus) and six Columbia black-tailed deer (Odocoileus hemionus columbianus) were held in pens and repeatedly anesthetized from April 2004 through June 2005 as part of an external parasite study. Deer were anesthetized using a combination of Telazol and xylazine hydrochloride (HCL) administered intramuscularly. Tolazoline HCL was slowly administered at 4 mg/kg intravenously to reverse the effects of xylazine with good results. For 17 of the 19 mule deer anesthesias in the fall of 2004, a mean dose of 7.3 mg/kg of intravenous tolazoline (range 6.1-8.4 mg/kg) was given by mistake. This paper describes clinical signs of apnea, muscle tensing, and fasciculations immediately following intravenous administration of tolazoline HCL in mule deer (O. hemionus) at 1.5-3 times the recommended dose. Mean dose for black-tailed deer during this time was 8.1 mg/kg (range 5.5-12.4 mg/kg) with no clinical signs as seen in the mule deer. Based on these findings, intravenous tolazoline use in mule deer is recommended at < or = 4 mg/kg.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
293490
2-Benzylimidazoline, 99%
C10H12N2