EMAIL THIS PAGE TO A FRIEND
Organic letters
Pentanidium-catalyzed enantioselective α-hydroxylation of oxindoles using molecular oxygen.
PMID 22946651
Abstract
Pentanidium-catalyzed α-hydroxylation of 3-substituted-2-oxindoles using molecular oxygen has been developed with good yields and enantioselectivities. This reaction does not require an additional reductant such as triethyl phosphite, which was typically added to reduce the peroxide intermediate. The reaction was demonstrated to consist of two-steps: an enantioselective formation of hydroperoxide oxindole and a kinetic resolution of the hydroperoxide oxindole via reduction with enolates generated from the oxindoles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9808
2-Oxindole, 97%
C8H7NO
T61204
Triethyl phosphite, 98%
C6H15O3P