EMAIL THIS PAGE TO A FRIEND
Organic letters
Regioselective catalytic hydroboration of propargylic species using Cu(I)-NHC complexes.
PMID 22946740
Abstract
The catalytic regioselective hydroboration of propargylic alcohols and ethers was investigated using NHC-CuCl. We observe that different NHC-CuCl complexes catalyze hydroborations of propargylic substrates with opposite regioselectivity. A 6-NHC-CuCl complex provides α-selectivity whereas β-selectivity is achieved using a 5-NHC-CuCl complex. The reaction tolerates a wide range of functional groups.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P50803
Propargyl alcohol, 99%
C3H4O